Learn More
CD4 Monoclonal Antibody (44.38), PE, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA181450
Additional Details : Gewicht : 0.01000kg
Beschreibung
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Sheep CD4 antibody, clone 44.38 recognizes the ovine CD4 cell surface glycoprotein, which is expressed by a subset of mature T lymphocytes.
CD4 Monoclonal antibody specifically detects CD4 in Goat, Ovine samples. It is validated for Flow Cytometry.Spezifikation
CD4 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P05542 | |
CD4 | |
Fetal thymocytes. | |
100 Tests | |
Primary | |
Goat, Ovine | |
Antibody | |
IgG2a |
Flow Cytometry | |
44.38 | |
PE | |
CD4 | |
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
Mouse | |
Protein A | |
RUO | |
102170124, 443509 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |