missing translation for 'onlineSavingsMsg'
Learn More

CD4 Rat anti-Mouse, FITC, Clone: GK1.5, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 561828

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15831379

  • 121.00 CHF / 1 Stück


Verfügbar ab
01-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The GK1.5 monoclonal antibody specifically binds to the mouse CD4 (L3T4) differentiation antigen. CD4 is expressed on most thymocytes, a subpopulation of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells), and a subset of NK-T cells. In addition, CD4 has also been reported to be detectable on pluripotent hematopoietic stem cells, bone marrow myeloid and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 has also been reported to be expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. The GK1.5 antibody reportedly blocks binding of the RM4-5 and H129.19, but not RM4-4 mouse CD4-specific antibodies.

Spezifikation

Spezifikation

CD4
GK1.5
FITC
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Rat
IgG2b, κ
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3
Affinity Purified
Mouse CTL clone V4
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.