missing translation for 'onlineSavingsMsg'
Learn More

CD4 Rat anti-Mouse, APC-H7, Clone: GK1.5, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 560246

Additional Details : Gewicht : 0.00800kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15820049

  • 181.00 CHF / 1 Stück
Verfügbar ab: 30-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The GK1.5 clone has been reported to react with the CD4 (L3T4) differentiation antigen expressed on most thymocytes, a subpopulation of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells), and a subset of NK-T cells. In addition, CD4 has also been reported to be detectable on pluripotent hematopoietic stem cells, bone marrow myeliod and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 has also been reported to be expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. GK1.5 mAb reportedly blocks binding of the RM4-5 (Mfr. No. 553046/553047) and H129.19 (Mfr. No. 553650/553651), but not RM4-4 (Mfr. No. 553055) antibodies.

Host Species: Rat
Clone: GK1.5
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse CTL clone V4

Flow Cytometry, Intracellular Staining

Spezifikation

Spezifikation

CD4
Monoclonal
0.2mg/mL
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
GK1.5
APC-H7
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Mouse CTL clone V4
25 μg
Immunology
Mouse
IgG2b κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.