Learn More
CD3e Hamster anti-Mouse, V450, Clone: 500A2, BD
Hamster Monoclonal Antibody
230.00 CHF - 465.00 CHF
Spezifikation
Antigen | CD3e |
---|---|
Klon | 500A2 |
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15840509
|
BD Biosciences
560804 |
25 μg |
230.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15820509
|
BD Biosciences
560801 |
0.1 mg |
465.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This antibody has been reported to react with the 25-kDa ε chain of the T-cell receptor-associated CD3 complex expressed on thymocytes, mature T lymphocytes, and NK-T cells on many mouse stains. Plate-bound and soluble hamster anti-mouse CD3e (clone 500A2) antibody has been reported to activate unprimed T cells in vitro. Activation of a mouse T-cell clone by soluble hamster anti-mouse CD3e (clone 500A2) can be blocked by Fab fragments of anti-CD4 antibody GK1.5, suggesting that the hamster anti-mouse CD3e (clone 500A2) antibody may bind an epitope of CD3e close to a site at which CD4 associates with the T-cell receptor. This hamster mAb to a mouse leukocyte antigen has been reported not to cross-react with rat leukocytes.
The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Spezifikation
CD3e | |
0.2mg/mL | |
Monoclonal | |
Hamster | |
Mouse | |
Mouse T-cell receptor | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
500A2 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2 κ | |
Affinity Purified | |
CD3ε chain |
For Research Use Only.