Learn More
CD3E, Human, Matched Antibody Pair (AP42), Abnova™
Artikelnummer. 16191834
Change view
Click to view available options
Menge:
1 Set
Packungsgröße:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16191834 1 Set 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16191834 Lieferant Abnova Lieferanten-Nr. H00000916AP42.1Set
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Includes
Capture antibody: mouse monoclonal anti-CD3E (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Spezifikation
Antigen CD3E
Anwendungen ELISA
Klassifikation Monoclonal/Polyclonal
Konjugat Biotin
Beschreibung This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD3E.
Gen CD3E
Gen-Alias FLJ18683/T3E/TCRE
Gensymbole CD3E
Wirtsspezies Mouse
Menge 1 Set
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 916
Enthält Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD3E (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Zielspezies Human
Inhalt und Lagerung Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid