missing translation for 'onlineSavingsMsg'
Learn More

CD38 Mouse anti-Human, PerCP-Cy5.5, Clone: HIT2, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 561106

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15830779

  • 301.00 CHF / 1 Stück
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The HIT2 monoclonal antibody specifically binds to CD38. The CD38 antigen is also known as T10, ADP-ribosyl cyclase 1, and cyclic ADP ribose hydrolase 1. CD38 is a 45 kDa type II single-chain transmembrane glycoprotein present on thymocytes, activated T cells and terminally differentiated B cells (plasma cells). CD38 is expressed by other cells including monocytes, macrophages, dendritic cells, NK cells, myeloid and erythroid precursors and some epithelial cells. The CD38 antigen acts as an ectoenzyme that catalyzes the synthesis and hydrolysis of a Ca++ mobilizing agent, cyclic ADP-ribose. This intracellular calcium plays an important role in cell signaling pathways leading to cellular growth, apoptosis, and differentiation. CD38 binds to CD31 and thus plays a role in lymphocyte adhesion to endothelial cells.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spezifikation

Spezifikation

CD38
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
HIT2
PerCP-Cyanine5.5
Mouse
25 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.