Learn More
CD38 Mouse anti-Human, PE, Clone: HB7, BD
Mouse Monoclonal Antibody
Marke:  BD Biosciences 342371
Additional Details : Gewicht : 0.03000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15825988
  • 850.00 CHF / 1 Stück
Verfügbar ab
27-05-2021
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD38, clone HB7, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with the BJAB cell line.
CD38 recognizes an integral membrane glycoprotein, Mr 45 kilodaltons (kd), with a protein core of 35 kd.
Spezifikation
Spezifikation
CD38
HB7
PE
Conjugated
Human BJAB B cell line
50 Tests
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Monoclonal
Flow Cytometry
12.5μg/mL
Phosphate buffered saline with BSA and 0.1% sodium azide.
Murine
IgG1, κ