Learn More
CD38 Mouse anti-Human, BUV395, Clone: HB7, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 563812
Additional Details : Gewicht : 0.01000kg
Beschreibung
The HB7 monoclonal antibody specifically binds to human CD38. CD38 is a type II transmembrane glycoprotein of 45 kDa with a protein core of 35 kDa. The CD38 antigen is expressed on essentially all pre-B lymphocytes, plasma cells, and thymocytes. It is also present on activated T lymphocytes, natural killer (NK) lymphocytes, myeloblasts, and erythroblasts. The antigen is expressed during the early stages of T- and B-lymphocyte differentiation, is lost during the intermediate stages of maturation, and then reappears during the final stages of maturation. The CD38 antigen is expressed on 90% of CD34+ cells, and is not expressed on pluripotent stem cells. Coexpression of CD38 antigen on CD34+ cells indicates lineage commitment of those cells. CD38 is a counter-receptor of CD31. It is also expressed in T- and B-acute lymphoblastic leukemia (ALL), Burkitt's lymphoma, multiple myeloma, acute myeloid leukemia (AML), and chronic lymphocytic leukemia (CLL).
The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon™ BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Spezifikation
CD38 | |
Monoclonal | |
Brilliant Ultraviolet 395 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human BJAB B cell line | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HB7 | |
T10; ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; OKT10 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only.