Learn More
CD3 Mouse anti-Human, PE, Clone: UCHT1, BD
Mouse Monoclonal Antibody
194.00 CHF - 1280.00 CHF
Spezifikation
Antigen | CD3 |
---|---|
Klon | UCHT1 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | PE |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15831359
|
BD Biosciences
561808 |
25 Tests |
194.00 CHF
1 Stück |
Verfügbar ab: 03-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15841359
|
BD Biosciences
561809 |
500 Tests |
1280.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.
Spezifikation
CD3 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
UCHT1 | |
Monoclonal | |
Mouse | |
Human | |
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.