Learn More
CD294 Rat anti-Human, Alexa Fluor 647, Clone: BM16, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561797
Additional Details : Gewicht : 0.01000kg
Beschreibung
The BM16 monoclonal antibody specifically binds to CD294. CD294 is encoded by PTGDR2 (Prostaglandin D2 receptor 2) and is also known as CRTH2 (chemoattractant receptor-homologous molecule expressed on Th2 cells), GPR44 (G protein-coupled receptor 44), DL1R, and DP2. CD294 is a member of the G protein-coupled leukocyte chemoattractant receptor family and it is expressed on Th2, but not Th1 type cells. CD294 is detectable on CD4+ T cells in fresh PBMC but not on B cells and NK cells. CD294 is also expressed on peripheral blood basophils and eosinophils, suggesting its involvement allergic reactions. Phenotypic analysis of CD4+ T cells expressing CRTH2 demonstrated that they were also CD45RA-negative and CD45RO+ and CD25+. These cells produce Th2- but little or no Th1-type cytokines upon stimulation with PMA and Ionomycin.
Spezifikation
CD294 | |
Monoclonal | |
0.2mg/mL | |
CRTH2; PTGDR2 ; Prostaglandin D2 receptor 2; DL1R; DP2; GPR44 | |
Rat | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
BM16 | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CRTH2 Transfected Cell Line | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.