Learn More
CD27 Mouse anti-Human, V450, Clone: M-T271, BD
Mouse Monoclonal Antibody
356.00 CHF - 790.00 CHF
Spezifikation
Antigen | CD27 |
---|---|
Klon | M-T271 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Violet Fluor 450 (V450) |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830219
|
BD Biosciences
560458 |
30 Tests |
356.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15840209
|
BD Biosciences
560448 |
120 Tests |
790.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Reacts with a disulphide-linked 110kDa dimer of two polypeptide chains. CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on the subset of human thymocytes and on the majority of mature T lymphocytes. CD27 is highly induced on T cells after TCR stimulation. CD27 may be involved in cellular interaction of T and B lymphocytes. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: M-T271
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD27 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
Human | |
Human T-CLL cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
M-T271 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.