Learn More
CD27 Mouse anti-Human, APC-H7, Clone: M-T271, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 560222
Additional Details : Gewicht : 0.01100kg
Beschreibung
Reacts with a disulphide-linked 110kDa dimer of two polypeptide chains. CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on the subset of human thymocytes and on the majority of mature T lymphocytes. CD27 is highly induced on T cells after TCR stimulation. CD27 may be involved in cellular interaction of T and B lymphocytes.
Host Species: Mouse
Clone: M-T271
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD27 | |
Monoclonal | |
APC-H7 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M-T271 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human T-CLL cells | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
For Research Use Only.