Learn More
CD25 Mouse anti-Human, BB515, Clone: 2A3, BD
Mouse Monoclonal Antibody
287.00 CHF - 585.00 CHF
Spezifikation
Antigen | CD25 |
---|---|
Klon | 2A3 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Brilliant Blue 515 (BB515) |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15833669
|
BD Biosciences
564468 |
25 Tests |
287.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15823669
|
BD Biosciences
564467 |
100 Tests |
585.00 CHF
1 Stück |
Verfügbar ab: 06-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The 2A3 monoclonal antibody specifically binds to human CD25, the low-affinity alpha subunit of the Interleukin-2 Receptor (IL- 2Rα). CD25 associates with CD122 (IL-2Rβ chain) and CD132 (common γchain or γc) to f orm the high-affinity signal-transducing IL-2R complex. CD25 is expressed by subsets of thymocytes and peripheral blood lymphocytes including CD4+CD25+ regulatory T cells and memory T cells. CD25 antigen density increases on activated T cells including phytohemagglutinin (PHA)-, concanavalin A (Con A)-, and CD3-activated T lymphocytes. High levels of CD25 can be expressed by T lymphocytes from mixed lymphocyte cultures and by human T-lymphocyte leukemia virus (HTLV)-infected T-lymphocyte leukemia lines, for example, HUT-102. CD25 can also be expressed by activated B cells and macrophages. Recombinant IL-2 blocks the binding of the 2A3 antibody to PHA-activated T lymphocytes.
The antibody was conjugated to BD Horizon™ BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Spezifikation
CD25 | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
RUO | |
Human Phytohemagglutinin-activated T Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2A3 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified | |
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55 |
For Research Use Only.