Learn More
P-Glycoprotein Monoclonal Antibody (UIC2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516959
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody does not react with mouse or rat. Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for immunohistochemical applications is human kidney. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD243, clone UIC2 recognizes an extracellular conformational epitope of CD243, also known as MDR1 (multi-drug resistance protein 1) and Pgp (P-glycoprotein), a multi pass transmembrane protein and member of the ABC transporter (ATP-binding cassette) family, containing two ABC transporter type 1 domains and two ABC transporter domains.
P-Glycoprotein Monoclonal antibody specifically detects P-Glycoprotein in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation.Spezifikation
P-Glycoprotein | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P08183 | |
ABCB1 | |
Mouse Balb/c 3T3 fibroblasts transfected with human CD243 cDNA. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
UIC2 | |
Unconjugated | |
ABCB1 | |
ABC superfamily; P-glycoprotein; ABC20; ABCB1; Abcb1a; Abcb1b; Abcb4; ATP binding cassette subfamily B member 1; ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette sub-family B member 1; ATP-binding cassette sub-family B member 1A; ATP-binding cassette sub-family B member 1B; ATP-binding cassette transporter subfamily B, member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette, subfamily B (MDR/TAP), member 1A; ATP-binding cassette, sub-family B (MDR/TAP), member 1A; ATP-binding cassette, subfamily B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, subfamily B, member 1A; ATP-binding cassette, subfamily B, member 1B; ATP-dependent translocase ABCB1; ATP-dependent translocase ABCB1; multidrug r | |
Mouse | |
Protein A | |
RUO | |
5243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |