missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GYPC Monoclonal Antibody (BRIC4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516592
Additional Details : Gewicht : 0.01000kg
Beschreibung
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
GYPC Monoclonal antibody specifically detects GYPC in Human samples. It is validated for Flow Cytometry, Hemagglutination Assay, Immunohistochemistry (Frozen), Western Blot.Spezifikation
GYPC | |
Monoclonal | |
0.5 mg/mL | |
TBS with ≤100mM glycine and <0.1% sodium azide | |
P04921 | |
GYPC | |
Human erythrocytes | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Hemagglutination Assay, Immunohistochemistry (Frozen), Western Blot | |
BRIC4 | |
Unconjugated | |
GYPC | |
0610037F22Rik; CD236; CD236R; GE; Gerbich antigen; GLPC; Glycoconnectin; glycophorin C; glycophorin C (Gerbich blood group); glycophorin-C; Glycophorin-D; glycoprotein beta; GPC; GPD; GPYD; GYPC; GYPD; MGC117309; MGC126191; MGC126192; PAS-2; PAS-2'; Sialoglycoprotein D | |
Mouse | |
Protein G | |
RUO | |
2995 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |