Learn More
CD235a Mouse anti-Human, PE, Clone: GA-R2 (HIR2), BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 340947
Additional Details : Gewicht : 0.03000kg
Beschreibung
Anti–glycophorin A reacts with a major sialoglycoprotein present on the human erythrocyte membrane. Glycophorin A is a transmembrane dimeric complex of 31 kilodaltons (kd) with its caboxyterminal ends extending into the cytoplasm of the red cell.
Spezifikation
CD235a | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
50 Tests | |
Cell Biology | |
Human | |
IgG2b κ |
Flow Cytometry | |
GA-R2 (HIR2) | |
PE | |
Mouse | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
For Research Use Only.