missing translation for 'onlineSavingsMsg'
Learn More

CD226 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531111

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 13435627

  • 674.00 CHF / 100 Mikroliter
Verfügbar ab: 18-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Rhesus Monkey (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CD226 Polyclonal antibody specifically detects CD226 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

CD226
Polyclonal
Unconjugated
Cd226
adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1
Rabbit
Antigen affinity chromatography
RUO
10666, 307199
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q15762
Cd226
Recombinant fragment corresponding to a region within amino acids 1 and 274 of Human CD226
100 μL
Primary
Human, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote