Learn More
IGF1R alpha Monoclonal Antibody (1H7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA181548
Additional Details : Gewicht : 0.01000kg
Beschreibung
This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. A suggested positive control for immunohistochemical applications is pancreas or placenta. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD221 antibody, clone 1H7 recognizes human CD221, a approximately 155 kDa receptor tyrosine kinase, also known as Insulin-like growth factor I receptor (IGF-I Receptor).
IGF1R alpha Monoclonal antibody specifically detects IGF1R alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IGF1R alpha | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P08069 | |
Igf1r | |
Purified human placental IGF-I receptor | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1H7 | |
Unconjugated | |
Igf1r | |
A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Mouse | |
Protein G | |
RUO | |
3480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |