Learn More
DC-SIGN Monoclonal Antibody (ER-TR9), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA140070
Additional Details : Gewicht : 0.01000kg
Beschreibung
MA1-40070 detects SIGN-R1 from mouse samples. MA1-40070 has been successfully used in flow cytometry, functional assays, immunohistochemistry (paraffin) and immunohistochemistry (frozen) applications. The MA1-40070 immunogen is mouse SIGN-R1.
DC-SIGN Monoclonal antibody specifically detects DC-SIGN in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Spezifikation
DC-SIGN | |
Monoclonal | |
Conc. not determined | |
antibody with cell culture media and 0.02% sodium azide | |
Q91ZX1 | |
Cd209a | |
Murine SIGN-R1. | |
1 mL | |
Primary | |
Mouse | |
Antibody | |
IgM |
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
ER-TR9 | |
Unconjugated | |
Cd209a | |
CD209; CD209 antigen; CD209 antigen-like protein A; CD209 molecule; Cd209a; CD209a antigen; CD209a molecule; Cd209d; CD209d antigen; CDSIGN; Cire; CLEC4L; Clec4m; C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; C-type lectin domain family 4, member M; Dcsign; DC-SIGN; DC-SIGN1; Dc-signr; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin; dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443; RGD1561104; SIGN-R1; SIGNR5 | |
Rat | |
purified | |
RUO | |
170786 | |
4° C | |
Liquid |