Learn More
CD192 Mouse anti-Human, Alexa Fluor 647, Clone: 48607, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 558406
Additional Details : Gewicht : 0.02000kg
Beschreibung
The 48607 monoclonal antibody specifically binds to CD192 which is also known as, Chemokine C-C motif receptor 2 (CCR2), Monocyte chemoattractant protein 1 receptor (MCP-1 receptor/MCP-1-R), or CKR2. CD192 is a seven-transmembrane, G-protein-coupled receptor that is the specific receptor for Monocyte chemoattractant protein 1 (MCP-1/CCL2), MCP-2/CCL8, MCP-3/CCL7, and MCP-4/CCL13. CD192 is expressed on monocytes, dendritic cells, NK cells, basophils, and activated T cells and B cells. CD192 exists in two forms, CD192A and CD192B. The two forms are derived from alternatively spliced variants of a single gene and differ at the C-terminal end. CD192 plays an important role in inflammatory responses.
Spezifikation
CD192 | |
Monoclonal | |
Alexa Fluor 647 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human hCCR2-transfected NS0 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
48607 | |
CCR2; MCP-1-R; CC-CKR-2; CMKBR2; CKR2; CCR2A; CCR2B; CKR2A; CKR2B | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2b |
For Research Use Only.