missing translation for 'onlineSavingsMsg'
Learn More

CD163 Monoclonal Antibody (GHI/61), FITC, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA517719

312.68 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15344001

  • 469.00 CHF / 100 Mikrogramm
Verfügbar ab: 18-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

MA5-17719 targets CD163/M130 in FACS applications and shows reactivity with Human samples. The MA5-17719 immunogen is mouse monoclonal antibodies raised against human CD163.

CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD163
Monoclonal
0.1 mg/mL
PBS with 4-5mg/mL BSA and 0.02% sodium azide
Q86VB7
CD163
Mouse monoclonal antibodies raised against human CD163.
100 μg
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
GHI/61
FITC
CD163
CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163
Mouse
Protein G
RUO
9332
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote