Learn More
CD16 Mouse anti-Human, PE-Cy7, Clone: 3G8, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 560716
Additional Details : Gewicht : 0.01000kg
Beschreibung
Reacts with the 50-65kDa transmembrane form of IgG Fc (FcγRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. CD16 antibody blocks the binding of soluble immune complexes to granulocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 3G8
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD16 | |
Monoclonal | |
PE-Cyanine7 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human polymorphonuclear leukocytes | |
50 Tests | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
3G8 | |
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.