missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD144 Mouse anti-Human, FITC, Clone: 55-7H1, BD
Mouse Monoclonal Antibody
229.00 CHF - 615.00 CHF
Spezifikation
Antigen | CD144 |
---|---|
Klon | 55-7H1 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | FITC |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830559
|
BD Biosciences
560874 |
25 Tests |
229.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15840179
|
BD Biosciences
560411 |
100 Tests |
615.00 CHF
1 Stück |
Verfügbar ab: 22-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The 55-7H1 antibody reacts with a calcium-independent epitope on cadherin-5, a member of the cadherin family of calcium-dependent adhesion molecules. Cadherin-5 is expressed on endothelial cells in vivo and in vitro. It may play a role in the organization of lateral endothelial junctions and in the control of permeability properties of vascular endothelium.
Host Species: Mouse
Clone: 55-7H1
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD144 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
55-7H1 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified | |
VE-cadherin; Cadherin-5; CDH5; Vascular endothelial cadherin |
For Research Use Only.