Learn More
CD144 Mouse anti-Human, FITC, Clone: 55-7H1, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 560874
Additional Details : Gewicht : 0.01000kg
Beschreibung
The 55-7H1 antibody reacts with a calcium-independent epitope on cadherin-5, a member of the cadherin family of calcium-dependent adhesion molecules. Cadherin-5 is expressed on endothelial cells in vivo and in vitro. It may play a role in the organization of lateral endothelial junctions and in the control of permeability properties of vascular endothelium.
Host Species: Mouse
Clone: 55-7H1
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD144 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
55-7H1 | |
VE-cadherin; Cadherin-5; CDH5; Vascular endothelial cadherin | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.