Learn More
CD140b Mouse anti-Human, Brilliant Violet 421, Clone: 28D4, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 564124
Beschreibung
Reacts with platelet derived growth factor (PDGF) receptor β , a 170-190 kDa single transmembrane glycoprotein expressed on fibroblasts, smooth muscle cells, glial cells and chondrocytes. PDGF receptors, α and β , are single glycoproteins with intracellular tyrosine kinase domains. They are structurally similar to M-CSF receptor and CD117 (c-kit). Their ligand, PDGF, is a mitogen for connective tissue cells and glial cells. PDGF plays a role in wound healing and acts as a chemoattractant for fibroblasts, smooth muscle cells, glial cells, monocytes and neutrophils. Functional PDGF is secreted as a disulfide-linked dimer of A and B chains (PDGF-AA, PDGF-BB or PDGF-AB). Binding of divalent PDGF induces receptor dimerization with three possible forms: αα , αβ , ββ . The PDGF receptor α β and does not cross-react with PDGFR α .
Spezifikation
| CD140b | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 28D4 | |
| PDGF Receptor β chain; PDGFRβ; PDGFRB; PDGFR1; PDGFR; PGFRB; JTK12 | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.