Learn More
CD14 Mouse anti-Human, V450, Clone: MΦP9, BD
Mouse Monoclonal Antibody
371.00 CHF - 790.00 CHF
Spezifikation
Antigen | CD14 |
---|---|
Klon | MΦP9 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Violet Fluor 450 (V450) |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15820129
|
BD Biosciences
560350 |
30 Tests |
371.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15810129
|
BD Biosciences
560349 |
120 Tests |
790.00 CHF
1 Stück |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Reacts with a 53-55kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. The purified antibody is suitable for staining acetone-fixed, frozen tissue sections. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: MφP9
Isotype: IgG2b, κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD14 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
Human | |
Human Monocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MΦP9 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified |
For Research Use Only.