missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Mouse anti-Human, PerCP-Cy5.5, Clone: M5E2, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561116
Additional Details : Gewicht : 0.01000kg
Beschreibung
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.Spezifikation
CD14 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CD14 Protein | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M5E2 | |
PerCP-Cyanine5.5 | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
IgG2a κ |
For Research Use Only.