Learn More
CD14 Mouse anti-Human, PE-Cy7, Clone: M5E2, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 557742
Additional Details : Gewicht : 0.01000kg
Beschreibung
Reacts with the 53-55kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Mouse
Clone: M5E2
Isotype: IgG2a, κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD14 | |
Monoclonal | |
PE-Cyanine7 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M5E2 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD14 Protein | |
100 Tests | |
Immunology | |
Canine, Human | |
IgG2a κ |
For Research Use Only.