Learn More
CD14 Mouse anti-Human, PE-Cy7, Clone: M5E2, BD
Mouse Monoclonal Antibody
298.00 CHF - 595.00 CHF
Spezifikation
Antigen | CD14 |
---|---|
Klon | M5E2 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | PE-Cyanine7 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830599
|
BD Biosciences
560919 |
25 Tests |
298.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15830999
|
BD Biosciences
561385 |
50 Tests |
530.00 CHF
1 Stück |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15819028
|
BD Biosciences
557742 |
100 Tests |
595.00 CHF
1 Stück |
Verfügbar ab: 10-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Reacts with the 53-55kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Mouse
Clone: M5E2
Isotype: IgG2a, κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD14 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
M5E2 | |
Monoclonal | |
Mouse | |
Immunology | |
Human CD14 Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.