Learn More
CD14 Mouse anti-Human, APC, Clone: M5E2, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555399
Additional Details : Gewicht : 0.02000kg
Beschreibung
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
Spezifikation
CD14 | |
Monoclonal | |
APC | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
M5E2 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD14 Protein | |
100 Tests | |
Immunology | |
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
IgG2a κ |
For Research Use Only.