Learn More
CD11b/Mac-1 Mouse anti-Human, APC, Clone: ICRF44, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561015
Additional Details : Gewicht : 0.01000kg
Beschreibung
Reacts with the 165kDa adhesion glycoprotein (Mac-1, integrin αM subunit) which associates with the 95kDa integrin β2 (CD18) to form the CD11b/CD18 complex. CD11b/Mac-1 is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and as a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits PMN aggregation in response to fMLP. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: ICRF44
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Spezifikation
CD11b | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human monocytes | |
25 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
ICRF44 | |
MAC-1A; Mac-1; ITGAM; Integrin alpha M; CR3A; CR-3 alpha; Mo1; SLEB6 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.