missing translation for 'onlineSavingsMsg'
Learn More

CD107a Mouse anti-Human, FITC, Clone: H4A3, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560949

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15830629

  • 132.00 CHF / 1 Stück


Verfügbar ab
27-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Reacts with the heavily glycosylated 110kDa lysosomal-associated membrane protein, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: H4A3
Isotype: IgG1 κ
Species Reactivity: Human

Intracellular Staining

Spezifikation

Spezifikation

CD107a
H4A3
LAMP1; LAMP-1; LAMPA; LGP120
Affinity Purified
Human Adult Adherent Peripheral Blood Cells
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.