missing translation for 'onlineSavingsMsg'
Learn More

CD107a Mouse anti-Human, APC, Clone: H4A3, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560664

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15830389

  • 443.00 CHF / 1 Stück
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Reacts with the heavily glycosylated 110kDa lysosomal-associated membrane protein, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini.

Host Species: Mouse
Clone: H4A3
Isotype: IgG1, κ
Cross Reactivity: Human

Flow Cytometry

Spezifikation

Spezifikation

CD107a
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Adult Adherent Peripheral Blood Cells
50 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
H4A3
LAMP1; LAMP-1; LAMPA; LGP120
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.