missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD106 Rat anti-Mouse, FITC, Clone: 429 (MVCAM.A), BD
Rat Monoclonal Antibody
319.00 CHF - 471.00 CHF
Spezifikation
Antigen | CD106 |
---|---|
Klon | 429 (MVCAM.A) |
Konzentration | 0.5mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15831229
|
BD Biosciences
561678 |
0.1 mg |
319.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817158
|
BD Biosciences
553332 |
0.5 mg |
471.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The 429 monoclonal antibody specifically binds to both the long (∼110 kDa) transmembrane-spanning form and the truncated (∼47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α;4 β; 1 integrin) and LPAM-1 ( α;4 β; 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.Spezifikation
CD106 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
Vcam-1; Vascular cell adhesion molecule 1; Vascular cell adhesion protein 1 |
429 (MVCAM.A) | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
Mouse preadipose cell line PA6 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.