missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD106 Rat anti-Mouse, FITC, Clone: 429 (MVCAM.A), BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561678
Additional Details : Gewicht : 0.01000kg
Beschreibung
The 429 monoclonal antibody specifically binds to both the long (∼110 kDa) transmembrane-spanning form and the truncated (∼47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α;4 β; 1 integrin) and LPAM-1 ( α;4 β; 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.Spezifikation
CD106 | |
Monoclonal | |
0.5mg/mL | |
Vcam-1; Vascular cell adhesion molecule 1; Vascular cell adhesion protein 1 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
429 (MVCAM.A) | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse preadipose cell line PA6 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2a κ |
For Research Use Only.