Learn More
CD100 Mouse, Unlabeled, Clone: 30, BD
Mouse Monoclonal Antibody
370.00 CHF - 730.00 CHF
Spezifikation
Antigen | CD100 |
---|---|
Klon | 30 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15825579
|
BD Biosciences
610670 |
50 μg |
370.00 CHF
1 Set |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15835579
|
BD Biosciences
610671 |
150 μg |
730.00 CHF
1 Set |
Verfügbar ab: 23-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
CD100 was originally described as a 150 kDa surface T-cell antigen induced after T-cell activation. Human CD100 exhibits widespread gene expression but is most prominent in peripheral lymphocytes, thymus, skeletal muscle, and brain. The full CD100 protein is 862 amino acids with homolgy to the semaphorin family of proteins at its extracellular domain followed by a short Ig-like domain. The short cytoplasmic domain contains a putative tyrosine phosphorylation site which may be the site for association with intracellular kinases. Cells that overexpress CD100 induce a rapid aggregation of B cells and increase the CD90L-B cell aggregation. Because semaphorins are axon-guiding molecules for the nervous system, it has been proposed that CD100 may have a guidance role in the immune system.
Spezifikation
CD100 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Semaphorin 4D; Sema 4D |
30 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.