missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD10 Monoclonal Antibody (RXB.14), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA112031
Additional Details : Gewicht : 0.01000kg
Beschreibung
CD10 Monoclonal antibody specifically detects CD10 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD10 | |
Monoclonal | |
Unconjugated | |
Mme | |
6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase | |
Mouse | |
Protein A/G | |
RUO | |
4311 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry | |
RXB.14 | |
PBS with 0.08% sodium azide | |
P08473 | |
Mme | |
White blood cells. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |