missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC90A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542641
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ELHQLKQQVM DEVIKVRTDT KLDFNLEKSR VKELYSLNEK KLLELRTEIV Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rat: 93%
CCDC90A Polyclonal antibody specifically detects CCDC90A in Human samples. It is validated for Western Blot.Spezifikation
CCDC90A | |
Polyclonal | |
Unconjugated | |
MCUR1 | |
6230416A05Rik; AU015498; AV136929; AW554392; C6orf79; C88263; Ccdc90a; coiled-coil domain containing 90A; coiled-coil domain-containing protein 90A, mitochondrial; FMP32; formin-like protein 16; MCU regulator 1; MCUR1; MCURI1; Mitochondrial calcium uniporter regulator 1; RGD1307673 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
63933 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96AQ8 | |
MCUR1 | |
Synthetic peptide directed towards the middle region of human CCDC90A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |