Learn More
δ-Catenin Mouse, Unlabeled, Clone: 30, BD
Mouse Monoclonal Antibody
370.00 CHF - 680.00 CHF
Spezifikation
Antigen | δ-Catenin |
---|---|
Klon | 30 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15815969
|
BD Biosciences
611536 |
50 μg |
370.00 CHF
1 Set |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15825969
|
BD Biosciences
611537 |
150 μg |
680.00 CHF
1 Set |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The catenins (α, β, γ, δ, p120[ctn]) are cytoplasmic proteins that are related to the Drosophila Armadillo protein. Catenins may have dual roles since they are components of cell-cell adherens junctions and can translocate to the nucleus after stimulation of the Wingless (Wnt-1 homolog) signaling pathway. α-Catenin has two subtypes: αE-Catenin, which is expressed ubiquitously, and αN-Catenin, which is expressed in the nervous system. β-Catenin binds to the cytoplasmic tail of E-Cadherin at adherens junctions and has been implicated in Wnt-1 signaling. γ-Catenin is associated with desmoglein in desmosomes and is closely related to β-Catenin. p120[ctn] is related to both β- and γ-catenin and is a substrate of tyrosine kinases localized at adherens junctions. δ-catenin was identified by its ability to bind the Alzheimer's disease-related protein, presenilin-1. It is most closely related to p120[ctn] and the desmosomal protein, p0071. It contains 10 Armadillo (Arm) repeats, as compared to the 13 Arm repeats found in β-Catenin. δ-Catenin is expressed at high levels in the developing nervous system, where it may be involved in neuronal progenitor cell migration and dendrite development.
Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse δ-Catenin aa. 85-194
Formula Weight [Chemical]: 160kDa
Immunofluorescence, Western Blotting
Spezifikation
δ-Catenin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
30 | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Rat | |
Mouse δ-Catenin aa. 85-194 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.