Learn More
CaM Kinase II Mouse, Unlabeled, Clone: 45, BD
Mouse Monoclonal Antibody
349.00 CHF - 680.00 CHF
Spezifikation
Antigen | CaM Kinase II |
---|---|
Klon | 45 |
Konzentration | 250μg/mL |
Anwendungen | In vitro Assay, Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15815869
|
BD Biosciences
611292 |
50 μg |
349.00 CHF
1 Set |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15825869
|
BD Biosciences
611293 |
150 μg |
680.00 CHF
1 Set |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Ca2+;/calmodulin-dependent protein kinase II (CaM kinase II) is a multifunctional Ser/Thr kinase that regulates a number of cellular functions in response to increased intracellular Ca2+;. CaM kinase II is widely distributed, but is predominantly expressed in brain. It is involved in the regulation of neuronal functions such as neurotransmitter synthesis, neurotransmitter release, long-term potentiation, and formation of spatial learning. Neuronal CaM kinase II contains heteromers of two major subunits, α and β, at a ratio of 2:1 and homomers of α subunits. Each subunit has N-terminal ATP-binding and catalytic/regulatory domains and a C-terminal association domain. The regulatory domain consists of the autoinhibitory and calmodulin-binding sites. Assembly of the association domains of multiple subunits positions the regulatory domains for intersubunit autophosphorylation. After binding Ca2+;/calmodulin, CaM kinase II undergoes rapid autophosphorylation of the α and β subunits, which results in a substantial increase in its affinity for Ca2+;/calmodulin.
Host Species: Mouse
Clone: 45
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat CaM Kinase IIα aa. 448-460
Formula Weight [Chemical]: 52kDa
Immunofluorescence, Immunohistochemistry, Western Blotting
Spezifikation
CaM Kinase II | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Ca2+/calmodulin-dependent protein kinase II |
45 | |
In vitro Assay, Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Rat CaM Kinase IIα aa. 448-460 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.