missing translation for 'onlineSavingsMsg'
Learn More

c-Jun Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA1833

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 11599100

  • 578.00 CHF / 200 Mikrogramm
Verfügbar ab: 29-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

PA1-833 detects cJun from mouse cells. PA1-833 has been successfully used in Western blot and ChIP procedures. By Western blot, this antibody detects an ∽40 kDa protein representing cJun in NIH 3T3 serum stimulated cells. A smaller band is also detected at ∽26 kDa possibly representing a proteolytic fragment of cJun. The PA1-833 immunogen is a synthetic peptide corresponding to residues T(91) P T P T Q F L C P K N V T D(105) of mouse cJun. Reconstitute in 200 μL PBS to create a stock of 1 mg/mL.

c-Jun Polyclonal antibody specifically detects c-Jun in Mouse samples. It is validated for ChIP Assay, Western Blot.
Spezifikation

Spezifikation

c-Jun
Polyclonal
Unconjugated
JUN
Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Rabbit
IgG fraction
RUO
16476
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
ChIP Assay, Western Blot
1 mg/mL
antibody with 0.05% sodium azide
P05627
JUN
Synthetic peptide corresponding to residues T(91) P T P T Q F L C P K N V T D(105) of mouse cJun.
200 μg
Primary
Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote