missing translation for 'onlineSavingsMsg'
Learn More

BiP/GRP78 Mouse, Unlabeled, Clone: 40, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610978

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15855739

  • 370.00 CHF / 1 Set
Verfügbar ab: 30-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Synthesis of nascent proteins occurs at sites on the endoplasmic reticulum (ER) called translocons. Translocon proteins form a pore in the membrane that allow passage of the newly synthesized protein from the ribosome into the ER lumen. As the nascent protein enters the lumen, it is bound by BiP ( bi nding p rotein), the major chaperone of the ER. This protein is identical to the 78kDa glucose regulated protein, GRP78. BiP binds short hydrophobic sequences of the emerging peptide and prevents denaturation or nonspecific aggregation. Hydrolysis of ATP by BiP results in the release of the nascent protein which quickly assumes its proper conformation. However, if folding is incorrect, BiP again binds the protein and prevents its exit from the ER. In addition, BiP binding is thought to enhance the movement of secretory polypeptides across the ER membrane, but it is not required for protein translocation. It is 60% identical to Hsp70 and is similarly increased by conditions that produce incorrectly folded proteins. Thus, BiP is a chaperone of the ER lumen that binds misfolded or unassembled secretory proteins and ensures proper movement of proteins from the ER to the Golgi apparatus.

Spezifikation

Spezifikation

BiP/GRP78
Monoclonal
250μg/mL
GRP78
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
40
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human BiP/GRP78 aa. 525-628
50 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG2a
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.