missing translation for 'onlineSavingsMsg'
Learn More

BAI1-associated protein 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Artikelnummer. 16112566
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16112566 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16112566 Lieferant Abnova Lieferanten-Nr. H00010458D01P.100ug

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit polyclonal antibody raised against a full-length human BAIAP2 protein.

The protein encoded by this gene has been identified as a brain-specific angiogenesis inhibitor (BAI1)-binding protein. This adaptor protein links membrane bound G-proteins to cytoplasmic effector proteins. This protein functions as an insulin receptor tyrosine kinase substrate and suggests a role for insulin in the central nervous system. It also associates with a downstream effector of Rho small G proteins, which is associated with the formation of stress fibers and cytokinesis. This protein is involved in lamellipodia and filopodia formation in motile cells and may affect neuronal growth-cone guidance. This protein has also been identified as interacting with the dentatorubral-pallidoluysian atrophy gene, which is associated with an autosomal dominant neurodegenerative disease. Alternative splicing results in multiple transcript variants encoding distinct isoforms

Sequence: MSLSRSEEMHRLTENVYKTIMEQFNPSLRNFIAMGKNYEKALAGVTYAAKGYFDALVKMGELASESQGSKELGDVLFQMAEVHRQIQNQLEEMLKSFHNELLTQLEQKVELDSRYLSAALKKYQTEQRSKGDALDKCQAELKKLRKKSQGSKNPQKYSDKELQYIDAISNKQGELENYVSDGYKTALTEERRRFCFLVEKQCAVAKNSAAYHSKGKELLAQKLPLWQQACADPSKIPERAVQLMQQVASNGATLPSALSASKSNLVISDPIPGAKPLPVPPELAPFVGRMSAQESTPIMNGVTGPDGEDYSPWADRKAAQPKSLSPPQSQSKLSDSYSNTLPVRKSVTPKNSYATTENKTLPRSSSMAAGLERNGRMRVKAIFSHAAGDNSTLLSFKEGDLITLLVPEARDGWHYGESEKTKMRGWFPFSYTRVLDSDGSDRLHMSLQQGKSSSTGNLLDKDDLAIPPPDYGAASRAFPAQTASGFKQRPYSVAVPAFSQGLDDYGARSMSRNPFAHVQLKPTVTNDRCDLSAQGPEGREHGDGSARTLAGR

Spezifikation

Antigen BAI1-associated protein 2
Anwendungen Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Beschreibung Rabbit polyclonal antibody raised against a full-length human BAIAP2 protein.
Zusammensetzung PBS with no preservative; pH 7.4
Gen BAIAP2
Gen-Zugriffsnummer NM_017451
Gen-Alias BAP2/IRSP53
Gensymbole BAIAP2
Wirtsspezies Rabbit
Immunogen BAIAP2 (NP_059345.1, 1 a.a. ∼ 552 a.a) full-length human protein.
Reinigungsverfahren Affinity Purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 10458
Zielspezies Human
Inhalt und Lagerung Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Produkttyp Antibody
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.