missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCL2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA547852
Additional Details : Gewicht : 0.00010kg
Beschreibung
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
ASCL2 Polyclonal antibody specifically detects ASCL2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ASCL2 | |
Polyclonal | |
Unconjugated | |
ASCL2 | |
2410083I15Rik; achaete scute-like protein 2; achaete-scute complex homolog 2; achaete-scute complex homolog 2 (Drosophila); achaete-scute complex homolog-like 2; achaete-scute complex-like 2; achaete-scute complex-like protein 2; achaete-scute family bHLH transcription factor 2; achaete-scute homolog 2; Ascl2; ASH2; ASH-2; bHLH transcription factor; Mash2; BHLHA45; Class A basic helix-loop-helix protein 45; H M bHLHa45; HASH2; long transient receptor potential-related channel 5; mammalian achaete scute homolog 2; mammalian achaete/scute homologue 2; MASH2; mASH-2 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
430 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q99929 | |
ASCL2 | |
E. coli-derived recombinant human ASCL2/Mash2 Met1-Ala49 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |