Learn More
Alkaline Phosphatase Mouse anti-Human, Unlabeled, Clone: B4-78, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561432
Additional Details : Gewicht : 0.01000kg
Beschreibung
The B4-78 monoclonal antibody reacts with the tissue-nonspecific isozyme of alkaline phosphatase. Alkaline phosphatases are membrane-bound glycoproteins. Four isozymes of alkaline phosphatase exist in humans: placental, placental-like, intestinal, and liver/bone/kidney. Liver/bone/kidney alkaline phosphatase is also known as tissue-nonspecific alkaline phosphatase (TNAP). Human embryonic stem cells and embryonic carcinoma cells express high levels of tissue-nonspecific alkaline phosphatase that decrease upon differentiation. Genetic and biochemical studies suggest that TNAP plays a role in skeletal mineralization.
Spezifikation
Alkaline Phosphatase | |
Monoclonal | |
0.5mg/mL | |
Bone/Kidney/Liver Alkaline Phosphatase, TNAP, TNSALP, AP-TNAP, ALPL | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
B4-78 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Bone Alkaline Phosphatase | |
0.1 mg | |
Primary | |
Store undiluted at 4°C |