missing translation for 'onlineSavingsMsg'
Learn More

6x-His Tag Monoclonal Antibody (HIS.H8), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA1213151MG

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15386261

  • 1612.00 CHF / 1 Milligramm
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

MA1-21315 may be stored at 4° C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20° C, avoiding freeze/thaw cycles.

6x-His Tag Monoclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

6x-His Tag
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
Mouse
Protein A
RUO
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Tag
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
HIS.H8
Unconjugated
6His tag; 6X His; His Tag; Histidine Tag
6x His synthetic peptide.
1 mg
Primary
Antibody
IgG2b
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote