Learn More
Annexin V Mouse anti-Human, CoraLite 594, Clone: 1E6A8, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech CL594-66245-100UL
Artikelnummer. 16820996
  • 423.57 CHF / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The Annexin V antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Annexin V. This antibody recognizes human antigen. The Annexin V antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Spezifikation
Spezifikation
Annexin V
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunofluorescence, Flow Cytometry (Intracellular Staining)
1E6A8
CoraLite 594
100 μL
308