Learn More
Annexin V Mouse anti-Human, CoraLite 594, Clone: 1E6A8, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-66245-100UL
Beschreibung
The Annexin V antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Annexin V. This antibody recognizes human antigen. The Annexin V antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Spezifikation
| Annexin V | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1E6A8 | |
| CoraLite 594 | |
| 100 μL | |
| 308 | |
| IgG1 |