missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11804-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.Spezifikation
Annexin A3 | |
Polyclonal | |
Unconjugated | |
ANXA3 | |
35 alpha calcimedin, Annexin 3, annexin A3, Annexin III, ANX3, ANXA3, Lipocortin III, PAP III | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11745, 25291, 306 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35639, P12429, P14669 | |
Anxa3 | |
Annexin A3 Fusion Protein Ag2381 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Liquid |