missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A11 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marke: Novus Biologicals NB300-875
This item is not returnable.
View return policy
Description
Annexin A11 Polyclonal specifically detects Annexin A11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Annexin A11 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| annexin A11, Annexin-11, ANX1156-kD, CAP50, CAP-50 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 311 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.2 - 1 ug/ml, Immunohistochemistry 3 - 5 ug/ml, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_001148 | |
| ANXA11 | |
| Peptide with sequence YPGYPPPPGGYPP-C corresponding to N-Terminus according to NP_001148.1, NP_665875.1, NP_665876.1. | |
| 0.1 mg | |
| Primary | |
| NP_001148.1; NP_665875.1 and NP_665876.1 are varients that encode the same protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction