missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AMPK alpha-1/beta-1/gamma-2, His Tag Recombinant Protein

Human AMPK alpha-1/beta-1/gamma-2, His Tag Recombinant Protein

Marke:  Invitrogen™ PV6240

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15438903

  • 8710.00 CHF / 1 Milligramm
Verfügbar ab: 05-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

AMPK is activated in response to stresses that deplete cellular ATP (low glucose, hypoxia and ischemia) and via signaling pathways in response to adiponectin, leptin and CAMKKβ. Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tags.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Spezifikation

Spezifikation

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
63.6/38/65kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
His Tag
50mM sodium phosphate pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol
1mg
aa 10-end
Recombinant
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote